Learn More
Sino Biological Recombinant Anti-Fetuin A/FETUA Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10318R00120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human Fetuin-A / AHSG (rh Fetuin-A; Catalog#10318-H08H; NP_001613.2; Met 1-Val 367).
Catalog No. 50-240-8356