Learn More
Sino Biological Anti-Fetuin A/FETUA Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10318MM0220
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Fetuin-A / AHSG (rhFetuin-A; Catalog#10318-H08H; NP_001613.2; Met 1-Val 367). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-7798