Learn More
Sino Biological Anti-Fetuin A/FETUA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 205452T02100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human Fetuin A / AHSG fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-2140