Learn More
Sino Biological Human AIF1 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 14227R03120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human IBA1 / AIF1 (rh IBA1 / AIF1; Catalog#14227-H07E; P55008-1; Met1-Pro147).
Catalog No. 50-237-1499