Learn More
Sino Biological Anti-Iba1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 14227T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Iba1 / AIF1 (rh Iba1 / AIF1; Catalog#14227-H07E; P55008-1; Met1-Pro147). Iba1 / AIF1 specific IgG was purified by Human Iba1 / AIF1 affinity chromatography.
Catalog No. 50-241-1575