Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human AKAP9 (aa 951-1098) Control Fragment Recombinant Protein

Catalog No. RP102407
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP102407 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP102407 Supplier Invitrogen™ Supplier No. RP102407
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (47%), Rat (47%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-110748. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The A-kinase anchor proteins are a group of structurally diverse proteins which have the common function of binding to the regulatory subunit of protein kinase A and confining the holoenzyme to discrete locations within the cell. This gene encodes a member of the AKAP family. Alternate splicing of this gene results in at least two isoforms that localize to the centrosome and the Golgi apparatus, and interact with numerous signaling proteins from multiple signal transduction pathways. These signaling proteins include type II protein kinase A, serine/threonine kinase protein kinase N, protein phosphatase 1, protein phosphatase 2a, protein kinase C-epsilon and phosphodiesterase 4D3.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q99996
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 10142
Name Human AKAP9 (aa 951-1098) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 5730481H23Rik; A kinase (PRKA) anchor protein (yotiao) 9; A kinase (PRKA) anchor protein 9; AKAP 120-like protein; AKAP 350; AKAP 450; AKAP350; AKAP450; AKAP9; AKAP-9; AKAP9-BRAF fusion protein; A-kinase anchor protein 350 kDa; A-kinase anchor protein 450 kDa; A-kinase anchor protein 9; A-kinase anchoring protein 9; AW545847; C79026; centrosome- and Golgi-localized PKN-associated protein; centrosome- and golgi-localized protein kinase N-associated protein; CG-NAP; G1-448-15; HYPERION; KIAA0803; kinase N-associated protein; LQT11; mei2-5; mKIAA0803; MU-RMS-40.16A; PPP1R45; PRKA9; Protein hyperion; protein kinase A anchoring protein 9; protein kinase A-anchoring protein 9; protein phosphatase 1, regulatory subunit 45; protein yotiao; repro12; YOTIAO
Common Name AKAP9
Gene Symbol AKAP9
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence EKLELSQRLSDLSEQLKQKHGEISFLNEEVKSLKQEKEQVSLRCRELEIIINHNRAENVQSCDTQVSSLLDGVVTMTSRGAEGSVSKVNKSFGEESKIMVEDKVSFENMTVGEESKQEQLILDHLPSVTKESSLRATQPSENDKLQKE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.