Learn More
Raybiotech Inc C-Series Human Acute Kidney Injury Antibody Array 1 Kit.
Supplier:  Raybiotech Inc AAHAKI12
Encompass_Preferred
Detects 20 Human Cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Membrane Antibody Array. Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). No dedicated equipment required. Compatible with any chemiluminescent imaging system. Proteins detected: TIM-1 (KIM-1), Albumin, Osteopontin (SPP1), TFF3 (Trefoil Factor 3), Beta-2 Microglobulin, Clusterin, CXCL16, Osteoactivin (GPNMB), FABP1, MCP-1 (CCL2), TNF RI (TNFRSF1A), Calbindin, IP-10 (CXCL10), Cystatin C, HGF, MIF, Lipocalin-2 (NGAL), TIMP-1, VCAM-1 (CD106), VEGF-A
Catalog No. 50-194-8456