Learn More
Sino Biological Phospho-Akt (Thr308) Antibody, Rabbit MAb
Supplier:  Sino Biological 110547R000220
Encompass_Preferred
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the residues around Thr308 of Human Phospho-Akt.
Catalog No. 50-240-8155