Learn More
Bioworld HUMAN AKT1 (RECOMBINANT)
Supplier:  Bioworld 220602371
Encompass_Preferred
Recombinant Human AKT1 Source: Baculovirus, SF9 insect cells. Description : Recombinant Human AKT1 produced in SF9 is a glycosylated, polypeptide chain containing 481 amino acids and having a molecular mass of 60 KD. AKT1 is purified...
Catalog No. 50-253-337