Learn More
Sino Biological Human AKT1S1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 109888T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human AKT1S1 / PRAS40, and purified by antigen affinity chromatography.
Catalog No. 50-237-3333