Learn More
Sino Biological Recombinant Human Albumin Protein, Amine-Labeled Biotinylated
Catalog No. 502992688
Encompass_Preferred
A DNA sequence encoding Human ALB (NP_000468.1)(Asp25-Leu609) was expressed. The purified protein was biotinylated in vitro.
Catalog No. 50-299-2688 Supplier Sino Biological Supplier No. 10968HNAY2B100
A DNA sequence encoding Human ALB (NP_000468.1)(Asp25-Leu609) was expressed. The purified protein was biotinylated in vitro.