Learn More
Sino Biological Recombinant Human Albumin Protein, Amine-Labeled Biotinylated
Catalog No. 502993247
Encompass_Preferred
A DNA sequence encoding Human ALB (NP_000468.1)(Asp25-Leu609) was expressed. The purified protein was biotinylated in vitro.
Catalog No. 50-299-3247 Supplier Sino Biological Supplier No. 10968HNAY2B20
A DNA sequence encoding Human ALB (NP_000468.1)(Asp25-Leu609) was expressed. The purified protein was biotinylated in vitro.