Learn More
Sino Biological Recombinant Anti-Albumin Antibody (HRP), Rabbit Monoclonal
Supplier:  Sino Biological 68001R101H100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified Human Serum Albumin and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-240-8172