Learn More
Sino Biological Anti-Albumin Antibody, Mouse Monoclonal
Supplier:  Sino Biological 68001MM05T20
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells.
Catalog No. 50-255-2924