Learn More
Biomatik Corporation Human Albumin (ALB) CLIA Kit
Supplier:  Biomatik Corporation EKU08653
Encompass_Preferred
Human Albumin (ALB) CLIA Kit for detecting ALB in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-400ug/ml. Sensitivity: 0.52ug/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P61586. Gene ID: 387. Target Synonyms: Serum albumin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7764