Learn More
Abcam Human Albumin ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1087881X96TESTS
Encompass_Preferred
Human Albumin ELISA kit is a sandwich ELISA used to quantify human albumin (ALB) with a sensitivity of 0.79 ng/mL - Broad sample compatibility – works with urine, saliva, milk, cerebrospinal fluid (CSF), cell culture supernatants, cell lysates, and tissue samples - Wide dynamic range – Detects albumin levels from 3.125–200 ng/mL - Efficient assay process – Total assay time of approximately 3 hours - Comprehensive kit components: Includes pre-coated 96-well microplate
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC0900357