Learn More
Abcam Human Albumin ELISA Kit, Fluorescent, 1 x 96 Tests
Supplier:  Abcam AB2293861X96TESTS
Encompass_Preferred
Human Albumin ELISA Kit, Fluorescent is a single-wash 90-min Simplestep used to quantify Human Albumin with a sensitivity of 0.26 ng/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Fluorescent Sandwich ELISA - 530/570/590 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2372232