Learn More
Rockland Immunochemicals Human Albumin Peroxidase 1 mg
SDP
Catalog No. 501943756
Encompass_Preferred
Human serum albumin is the most abundant protein in human blood plasma. It is produced in the liver. Albumin constitutes about half of the blood serum protein. It is soluble and monomeric. Albumin transports hormones fatty acids and other compounds buffers pH and maintains osmotic pressure among other functions. Human Albumin is ideal for investigators involved in serum protein component research.
Catalog No. 50-194-3756 Supplier Rockland Immunochemicals Supplier No. 0090333
Human serum albumin is the most abundant protein in human blood plasma. It is produced in the liver. Albumin constitutes about half of the blood serum protein. It is soluble and monomeric. Albumin transports hormones fatty acids and other compounds buffers pH and maintains osmotic pressure among other functions. Human Albumin is ideal for investigators involved in serum protein component research.