Learn More
Sino Biological Anti-CD166/ALCAM Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10045RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD166 / ALCAM (rh CD166 / ALCAM; Catalog#10045-H08H; NP_001618.2; Met 1-Ala 526). CD166 / ALCAM specific IgG was purified by Human CD166 / ALCAM affinity chromatography.
Catalog No. 50-240-8468