Learn More
Sino Biological Recombinant Human CD166/ALCAM Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 10045H02H100
Encompass_Preferred
A DNA sequence encoding the human ALCAM (NP_001618.2) (Met1-Ala526) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-5517