Learn More
Biomatik Corporation Human Aldehyde Dehydrogenase 1 Family, Member L1 (ALDH1L1) ELISA Kit
Supplier:  Biomatik Corporation EKU11766
Encompass_Preferred
Human Aldehyde Dehydrogenase 1 Family, Member L1 (ALDH1L1) ELISA Kit for detecting ALDH1L1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O75891. Gene ID: 10840. Target Synonyms: FTHFD; 10-fTHF; 10-Formyltetrahydrofolate Dehydrogenase; Cytosolic 10-formyltetrahydrofolate dehydrogenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8087