Learn More
American Research Products Inc Human aldehyde dehydrogenase
Catalog No. 5000329533
Encompass
Human aldehyde dehydrogenase ALDH ELISA Kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-29533 Supplier American Research Products Inc Supplier No. CSBE11854H
Human aldehyde dehydrogenase ALDH ELISA Kit from CUSABIO Detection Range 78 pg/mL-5000 pg/mL Sensitivity 19 5 pg/mL Sample serum plasma tissue homogenates Method Sandwich