Learn More
Biomatik Corporation Human Aldehyde Dehydrogenase, Mitochondrial (ALDH2) ELISA Kit
Supplier:  Biomatik Corporation EKU02227
Encompass_Preferred
Human Aldehyde Dehydrogenase, Mitochondrial (ALDH2) ELISA Kit for detecting ALDH2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P05091. Gene ID: 217. Target Synonyms: ALDM; ALDH-E2; ALDHI; Aldehyde Dehydrogenase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7007