Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human ALDH18A1 (aa 97-245) Control Fragment Recombinant Protein

Catalog No. RP89746
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP89746 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP89746 Supplier Invitrogen™ Supplier No. RP89746
  • $283.00
    $212.25 / Each of 1

    Save $70.75

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52546. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene is a member of the aldehyde dehydrogenase family and encodes a bifunctional ATP- and NADPH-dependent mitochondrial enzyme with both gamma-glutamyl kinase and gamma-glutamyl phosphate reductase activities. The encoded protein catalyzes the reduction of glutamate to delta1-pyrroline-5-carboxylate, a critical step in the de novo biosynthesis of proline, ornithine and arginine. Mutations in this gene lead to hyperammonemia, hypoornithinemia, hypocitrullinemia, hypoargininemia and hypoprolinemia and may be associated with neurodegeneration, cataracts and connective tissue diseases. Alternatively spliced transcript variants, encoding different isoforms, have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P54886
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 5832
Name Human ALDH18A1 (aa 97-245) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 2810433K04Rik; ADCL3; AI429789; aldehyde dehydrogenase 18 family member A1; aldehyde dehydrogenase 18 family, member A1; aldehyde dehydrogenase 18A1; aldehyde dehydrogenase family 18 member A1; aldh18a1; ARCL3A; cb842; cb899; delta1-pyrroline-5-carboxlate synthetase; delta-1-pyrroline-5-carboxylate synthase; delta-1-pyrroline-5-carboxylate synthetase; Gamma-glutamyl kinase; Gamma-glutamyl phosphate reductase; GK; Glutamate 5-kinase; glutamate gamma-semialdehyde synthetase; Glutamate-5-semialdehyde dehydrogenase; Glutamyl-gamma-semialdehyde dehydrogenase; GPR; GSAS; MGC117316; P5CS; Pycs; pyrroline-5-carboxylate synthetase; pyrroline-5-carboxylate synthetase (glutamate gamma-semialdehyde synthetase); sb:cb881; similar to pyrroline-5-carboxylate synthetase isoform 1; SPG9A; SPG9B; wu:fa91f10; wu:fi05f11; wu:fi17d12
Common Name ALDH18A1
Gene Symbol ALDH18A1
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SIVEQVSVLQNQGREMMLVTSGAVAFGKQRLRHEILLSQSVRQALHSGQNQLKEMAIPVLEARACAAAGQSGLMALYEAMFTQYSICAAQILVTNLDFHDEQKRRNLNGTLHELLRMNIVPIVNTNDAVVPPAEPNSDLQGVNVISVKD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.