Learn More
Raybiotech Inc Human ALDH1A1 ELISA Kit
Supplier:  Raybiotech Inc ELHALDH1A11
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.4 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA pair antibody detects Human ALDH1A1
Catalog No. 50-194-8965