Learn More
Sino Biological Human ALDH1A1 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 11388MM0320
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ALDH1 (rh ALDH1; Catalog#11388-H07E; NP_000680.2; Ser 2-Ser 501). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2003