Learn More
Sino Biological Anti-ALDH1A1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11388RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ALDH1 (rh ALDH1; Catalog#11388-H07E; NP_000680.2; Ser 2-Ser 501). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8367