Learn More
Sino Biological Anti-ALDH9A1/ALDH9 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 102538T36100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ALDH9A1/ALDH9, and purified by antigen affinity chromatography.
Catalog No. 50-240-8140