Learn More
Sino Biological Recombinant Human ALK-1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10066H08H1
Encompass_Preferred
A DNA sequence encoding the the extracellular domain of human ALK1 (NP_000011.2) (Met 1-Gln 118) was fused with a polyhistide tag at the C-terminus.
Catalog No. 50-224-6661