Learn More
Sino Biological Recombinant Human ACVR1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10227H08B1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Val 124) of human ALK2 (Q04771) (Met 1-Val 124) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6813