Learn More
Sino Biological Recombinant Human ALK4/ACVR1B Protein (His Tag), HPLC-verified 1 mg
Supplier:  Sino Biological 10583H08H1
Encompass_Preferred
A DNA sequence encoding the human ACVR1B (NP_004293.1) extracellular domain (Met 1-Glu 126) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-224-7124