Learn More
Abcam Human Alkaline Phosphatase ELISA Kit, 96TEST
Supplier:  Abcam AB28524996TEST
Encompass_Preferred
Human Alkaline Phosphatase ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Alkaline Phosphatase in Human in Plasma, Tissue Homogenate, Serum, Other biological fluids samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-221-5499