Learn More
New England Biolabs, Inc. Human Alkyladenine Glycosylase (hAAG) – 500 units
SDP
Catalog No. 504362
Encompass
hAAG excises alkylated and oxidative DNA damaged sites and catalyzes the hydrolysis of the N-glycosidic bond to release the damaged base.
Catalog No. 50-436-2 Supplier New England Biolabs, Inc. Supplier No. M0313S
hAAG excises alkylated and oxidative DNA damaged sites and catalyzes the hydrolysis of the N-glycosidic bond to release the damaged base.