Learn More
Sino Biological Human ALOX15B PAb 100ul (Sino Biological)
Supplier:  Sino Biological 14564RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ALOX15B / 15 Lipoxygenase 2 (rh ALOX15B; Catalog#14564-H20B; AAH35217.1; Met1-Ile676). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-237-2781