Learn More
BIOHIPPO Human ALP Microsample
SDP
Catalog No. 5000314264
Encompass
Human ALP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ALP in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 067 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ALP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ALP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ALP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14264 Supplier BIOHIPPO Supplier No. BHE15212154
Human ALP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ALP in serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 067 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ALP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ALP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ALP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color