Learn More
Sino Biological Recombinant Human ALPPL2/ALPG Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 18763H08H1
Encompass_Preferred
A DNA sequence encoding the Human ALPG (P10696) (Met1-Asp503) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-255-3999