Learn More
Bioworld HUMAN ALPHA-1 ANTICHYMOTRYPSIN
Supplier:  Bioworld 220602411
Encompass_Preferred
Human Alpha-1 AntiChymotrypsin (ACT) Source: Human Plasma Description : Alpha 1 ACT is an early-stage acute-phase plasma protein and a serpin that preferentially inactivates chymotrypsin, cathepsin G, and chymase. Alpha-1- ACT, a ser...
Catalog No. 50-253-341