Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human alpha-1 PDX Recombinant Protein

Catalog No. RP070
Change view
Click to view available options
Quantity:
2.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP070 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP070 Supplier Invitrogen™ Supplier No. RP070
Add to Cart
Add to Cart

Recombinant Protein

RP-070 was obtained from E. coli transformed with the alpha1-PDX gene. RP-070 can be used in furin inhibition assays. For furin inhibition assays, cells should be treated with 8.0-20 μM alpha1-PDX for 1 hour (see above references). For in vitro assays, 0.1-2.0 μM alpha1-PDX should be used. Purity: >60% (most lots are 80-95%) NOTE: Freezing this product may result in damage of the protein. NOTE: Once rehydrated, this protein retains optimal reactivity for 2-4 days at 4 degrees C. Longer storage time can be achieved by storing in a serum rich medium. RP-070 can be used for protein biochemical, intracellular trafficking, and protein kinetic assays. Molecular weight is approximately 46.7 kDa.

Alpha1-PDX is a potent and selective inhibitor of furin (Ki = 0.6 nM). Alpha1-PDX was engineered by the construction of a variant of alpha1-antitrypsin that contains a minimal furin consensus sequence, RXXR. Cellular expression of alpha1-PDX effectively blocks the furin-dependent processing of protein precursors. In addition, extracellular application of alpha1-PDX can also block the processing of furin substrates. Alpha1-PDX has been shown to inhibit the processing of biologically significant proteins including many viral envelope glycoproteins, bacterial toxins, receptors and growth factors.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 2.5 mg/mL
For Use With (Application) Control, Inhibition Assay
Formulation PBS with no preservative; pH 7.2
Molecular Weight (g/mol) 46.7 kDa
Name Human alpha-1 PDX
pH Range 7.2
Quantity 2.5 mg
Source E. coli
Gene Alias alpha1 PDX; Pyruvate Dehydrogenase Complex Component-X
Common Name alpha-1 PDX
Biological Activity For furin inhibition assays, cells should be treated with 8.0 - 20 μM human alpha-1 PDX recombinant protein for 1 hour. For in vitro assays, 0.1 - 2.0 μM human alpha-1 PDX recombinant protein should be used.
Conjugate Unconjugated
Species E. coli
Recombinant Recombinant
Content And Storage 4°C, do not freeze
Expression System E. coli
Form Lyophilized
Protein Subtype Recombinant
Purity or Quality Grade >80%
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.