Learn More
Immunology Consultants Human Alpha 1-Microglobulin ELISA Kit
SDP
Supplier:  Immunology Consultants E80A1M
Encompass_Preferred
The Human Alpha 1-Microglobulin (A1M) test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring A1M in human biological samples.
Catalog No. 50-240-6633