Learn More
Biomatik Corporation Human Alpha 2-Antiplasmin (a2PI) CLIA Kit
Supplier:  Biomatik Corporation EKU11642
Encompass_Preferred
Human Alpha 2-Antiplasmin (a2PI) CLIA Kit for detecting a2PI in plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 1.56-100ng/ml. Sensitivity: 0.71ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P08697. Gene ID: 5345. Target Synonyms: SERPINF2; AAP; A2-PI; API; PLI; A2AP; Alpha-2-Plasmin Inhibitor; Serpin Peptidase Inhibitor Clade F Member 2; Pigment Epithelium Derived Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8110