Learn More
Invitrogen™ Human Alpha-Galactosidase A/GLA ELISA Kit
Description
Human Alpha-Galactosidase A/GLA quantitates human Alpha-Galactosidase A/GLA in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Alpha-Galactosidase A/GLA.
Specifications
Specifications
| Accession Number | P06280 |
| Assay Range | 45-10000 pg/mL |
| Assay Sensitivity | 45 pg/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | Agalsidase; agalsidase alfa; Ags; Alpha-D-galactosidase A; alpha-D-galactoside galactohydrolase; alpha-D-galactoside galactohydrolase 1; alpha-gal A; alpha-galactosidase A; GALA; galactosidase alpha; galactosidase, alpha; GLA; MelibiaseAgalsidase; agalsidase alfa; Ags; Alpha-D-galactosidase A; alpha-D-galactoside galactohydrolase; alpha-D-galactoside galactohydrolase 1; alpha-gal A; alpha-galactosidase A; GALA; galactosidase alpha; galactosidase, alpha; GLA; Melibiase; |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.