Learn More
ACROBiosystems Human Alpha-Synuclein, Tag Free (ALN-H5214) is expressed from E.coli cells. It contains AA Met 1 - Ala 140 (Accession # P37840-1).
Supplier:  ACROBiosystems ALNH5214200UG
Encompass_Preferred
Alpha-synuclein is a neuronal protein that plays several roles in synaptic activity such as regulation of synaptic vesicle trafficking and subsequent neurotransmitter release. It acts also as a molecular chaperone in its multimeric membrane-bound state, assisting in the folding of synaptic fusion components called SNAREs (Soluble NSF Attachment Protein REceptors) at presynaptic plasma membrane in conjunction with cysteine string protein-alpha/DNAJC5. Abnormalities in alpha-synuclein are implicated in the pathogenesis of Parkinson's disease (PD). Alpha-synuclein is present in Lewy-bodies, the neuropathological hallmark of PD, and the protein and its aggregation have been widely linked to neurotoxic pathways that ultimately lead to neurodegeneration.
Catalog No. 50-225-4135