Learn More
Enzyme Research Laboratories Inc Human a-Thrombin (Factor IIa),1000 U
Catalog No. NC9872266
Encompass
Please use this part # if ordering a quantity of 1-9. If ordering qty 10 or more please use Fisher part # NC0704726.
Human a-Thrombin (Factor IIa),1000 U,Buffer Composition = 50 mM Sodium Citrate,0.2 M NaCl,0.1% PEG-8000,pH 6.5,Extinction Coefficient (1%) = 18.3,Molecular Weight = 37,000 daltons,Prepared from human Prothrombin by activation with Factor Xa
Catalog No. NC9872266 Supplier Enzyme Research Laboratories Inc Supplier No. HT 1002A
Please use this part # if ordering a quantity of 1-9. If ordering qty 10 or more please use Fisher part # NC0704726.
Human a-Thrombin (Factor IIa),1000 U,Buffer Composition = 50 mM Sodium Citrate,0.2 M NaCl,0.1% PEG-8000,pH 6.5,Extinction Coefficient (1%) = 18.3,Molecular Weight = 37,000 daltons,Prepared from human Prothrombin by activation with Factor Xa