Learn More
Haematologic Technologies Inc HUMAN ALPHA THROMBIN 100UG
Supplier:  Haematologic Technologies Inc HCT0020
Encompass_Preferred
Coagulation Protein, Human alpha-Thrombin, 100 micrograms, Native enzyme, Mode of action: Serine protease which cleaves fibrinogen to form fibrin, Useful for cleavage of fusion proteins, Research use only, 50% (v/v) glyercol/water, store at -20 Celsius
Catalog No. 50-883-435