Learn More
Haematologic Technologies Inc HUMAN ALPHA THROMBIN 1MG
Supplier:  Haematologic Technologies Inc HCT00201MG
Encompass_Preferred
Coagulation Protein, Human alpha-Thrombin, 1 milligram, Native enzyme, Mode of action: Serine protease which cleaves fibrinogen to form fibrin, Research use only, 50% (v/v) glyercol/water, store at -20 Celsius
Catalog No. 50-883-394