Learn More
Biomatik Corporation Human Alpha-1-B-Glycoprotein (a1BG) ELISA Kit
Supplier:  Biomatik Corporation EKU02267
Encompass_Preferred
Human Alpha-1-B-Glycoprotein (a1BG) ELISA Kit for detecting a1BG in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.147ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04217. Gene ID: 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5118