Learn More
Biomatik Corporation Human Alpha-1-Microglobulin (a1M) CLIA Kit
Supplier:  Biomatik Corporation EKU08860
Encompass_Preferred
Human Alpha-1-Microglobulin (a1M) CLIA Kit for detecting a1M in serum, plasma, urine, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.36pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P02760. Gene ID: 259. Target Synonyms: AMBP; UTI; HCP; EDC1; HI30; IATIL; ITILC; ITI; ITIL; Alpha 1 Microglobulin/Bikunin Precursor; Growth-inhibiting protein 19; Uristatin; Uronic-Acid-Rich Protein; Trypstatin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8611