Learn More
Biomatik Corporation Human Alpha-1-Microglobulin/Bikunin Precursor, Ambp Elisa Kit
Supplier:  Biomatik Corporation EKC31135
Encompass_Preferred
Human Alpha-1-Microglobulin/Bikunin Precursor, Ambp Elisa Kit for detecting Alpha-1-Microglobulin/Bikunin Precursor in serum, plasma, urine, tissue homogenates. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.039ng/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 1-5h. Accession Number: P02760; AMBP. Target Synonyms: A1M; Alpha 1 microglobulin/bikunin precursor; Alpha 1 microglycoprotein; Alpha-1 microglycoprotein; Alpha-1-microglobulin; AMBP; AMBP_HUMAN; Bikunin; Complex-forming glycoprotein heterogeneous in charge; EDC1; Growth inhibiting protein 19; HCP; HI-30; HI30; IATIL; ITI; ITI-LC; ITIL; Microglobulin (alpha 1); Protein AMBP; Protein HC; Trypstatin; Uristatin; Uronic-acid-rich protein; UTI. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9198