Learn More
Biomatik Corporation Human Alpha-2-Glycoprotein 1, Zinc Binding (aZGP1) CLIA Kit
Supplier:  Biomatik Corporation EKU09909
Encompass_Preferred
Human Alpha-2-Glycoprotein 1, Zinc Binding (aZGP1) CLIA Kit for detecting aZGP1 in serum, plasma, urine, saliva, seminal plasma, sweat, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.82-600ng/ml. Sensitivity: 0.35ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P25311. Gene ID: 563. Target Synonyms: ZA2G; ZAG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4793