Learn More
Biomatik Corporation Human Alpha-2-Macroglobulin (a2M) CLIA Kit
Supplier:  Biomatik Corporation EKU08930
Encompass_Preferred
Human Alpha-2-Macroglobulin (a2M) CLIA Kit for detecting a2M in serum, plasma, urine, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.14-100ng/ml. Sensitivity: 0.05ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P01023. Gene ID: 2. Target Synonyms: A2MG; A2mg; CPAMD5; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5552